missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-7035257
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
603122-84-5
C8H8BFO4
197.96
MFCD08436034
BKWRLCIYMAYFPA-UHFFFAOYSA-N
2-fluoro-4-methoxycarbonyl phenylboronic acid, 2-fluoro-4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonyl-2-fluorophenylboronic acid, 2-fluoro-4-methoxycarbonyl phenyl boronic acid, o-fluoro p-methoxy carbonyl phenyl boronic acid, pubchem17434, acmc-209mj4, 2-fluoro-4-methoxycarbonyl phenylboronicacid, 2-fluoro-4-methoxycarbonyl-phenylboronic acid
24824345
(2-fluoro-4-methoxycarbonylphenyl)boronic acid
COC(=O)C1=CC(F)=C(C=C1)B(O)O
This item is not returnable. View return policy
603122-84-5
C8H8BFO4
197.96
MFCD08436034
BKWRLCIYMAYFPA-UHFFFAOYSA-N
2-fluoro-4-methoxycarbonyl phenylboronic acid, 2-fluoro-4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonyl-2-fluorophenylboronic acid, 2-fluoro-4-methoxycarbonyl phenyl boronic acid, o-fluoro p-methoxy carbonyl phenyl boronic acid, pubchem17434, acmc-209mj4, 2-fluoro-4-methoxycarbonyl phenylboronicacid, 2-fluoro-4-methoxycarbonyl-phenylboronic acid
24824345
(2-fluoro-4-methoxycarbonylphenyl)boronic acid
COC(=O)C1=CC(F)=C(C=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 173°C to 175°C
Quantity 250 mg
Formula Weight 197.96
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'