missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-nitro-6-methylpyridine, 98%, Thermo Scientific™

Catalog Number 15471149
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
19346-45-3
C6H5FN2O2
156.116
MFCD03095067
BCCWOQXQVNCXKC-UHFFFAOYSA-N
2-fluoro-3-nitro-6-picoline, 2-fluoro-3-nitro-6-methylpyridine, 2-fluoro-6-methyl-3-nitro-pyridine, 2-fluoro-6-methyl-3-nitro pyridine, pyridine, 2-fluoro-6-methyl-3-nitro, pubchem6309, abbypharma ap-30-0928, 6-fluoro-5-nitro-2-picoline, 2-fluoranyl-6-methyl-3-nitro-pyridine
2783178
2-fluoro-6-methyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])F
This item is not returnable. View return policy
19346-45-3
C6H5FN2O2
156.116
MFCD03095067
BCCWOQXQVNCXKC-UHFFFAOYSA-N
2-fluoro-3-nitro-6-picoline, 2-fluoro-3-nitro-6-methylpyridine, 2-fluoro-6-methyl-3-nitro-pyridine, 2-fluoro-6-methyl-3-nitro pyridine, pyridine, 2-fluoro-6-methyl-3-nitro, pubchem6309, abbypharma ap-30-0928, 6-fluoro-5-nitro-2-picoline, 2-fluoranyl-6-methyl-3-nitro-pyridine
2783178
2-fluoro-6-methyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
6-Methyl-3-nitro-2-(2-phenylhydrazinyl)pyridine (MNPHP) was synthesized from 2-fluoro-3-nitro-6-methylpyridine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 30°C
Color White
Quantity 250 mg
Beilstein 1638807
Solubility Information Slightly soluble in water.
Formula Weight 156.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Fluoro-3-nitro-6-methylpyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'