missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-3-methylbenzeneboronic acid pinacol ester, 97%

Brand:  Alfa Aesar™ H56391.MD

Additional Details : CAS Number : 1192548-08-5 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15472666

  • 1379.72 DKK / 250mg


Estimated Shipment
01-09-2020
Add to basket

Specifications

Chemical Identifiers

1192548-08-5
236.093
WJYURBPLHZZUGJ-UHFFFAOYSA-N
53217120
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C)F
C13H18BFO2
MFCD18384075
2-2-fluoro-3-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-3-methylphenylboronic acid,pinacol ester, 2-fluoro-3-methylphenylboronic acid, pinacol ester
2-(2-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

2-Fluoro-3-methylbenzeneboronic acid pinacol ester
1192548-08-5
MFCD18384075
WJYURBPLHZZUGJ-UHFFFAOYSA-N
2-(2-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
53217120
Solid
250mg
C13H18BFO2
2-2-fluoro-3-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-3-methylphenylboronic acid,pinacol ester, 2-fluoro-3-methylphenylboronic acid, pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C)F
236.093
236.09
97%
Videos
SDS Documents