missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-2'-deoxyinosine, 99%, Thermo Scientific Chemicals

Catalog Number p-7043043
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
80049-87-2
C10H11FN4O4
270.22
MFCD09750860
NRVOTDBYJXFINS-GPGUAWMRNA-N
196148
OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=NC2=C1N=CNC2=O
This item is not returnable. View return policy
80049-87-2
C10H11FN4O4
270.22
MFCD09750860
NRVOTDBYJXFINS-GPGUAWMRNA-N
196148
OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=NC2=C1N=CNC2=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Synonym 2'-fluoro-2'-deoxyinosine, 2'-deoxy-2'-fluoroinosine, inosine,2'-deoxy-2'-fluoro, 9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2'-dfin, 9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one, 2'-deoxy-2'-fluoro-inosine, inosine, 2'-deoxy-2'-fluoro, 9-2-deoxy-2-fluoro-beta-d-ribofuranosyl hypoxanthine, 9-2-deoxy-2-fluoro-beta-d-ribofuranosyl-9h-purin-6 1h-one
IUPAC Name 9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Formula Weight 270.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Fluoro-2'-deoxyinosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'