Få mere at vide
2-Fluoro-1-naphthalenecarboxaldehyde, TRC
Artikelnummer. 30240933
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30240933 250 mg -
30240731 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30240933 Leverandør Toronto Research Chemicals Leverandørnr. TRCF593795250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 82128-49-2
Molekylær formel C11 H7 F O
Molekylvægt (g/mol) 174.171
Synonym 2-Fluoro-1-naphthaldehyde
IUPAC navn 2-fluoronaphthalene-1-carbaldehyde
SMIL Fc1ccc2ccccc2c1C=O
Tekniske data
Kemisk navn eller materiale 2-Fluoro-1-naphthalenecarboxaldehyde
Mængde 250 mg
InChI formel InChI=1S/C11H7FO/c12-11-6-5-8-3-1-2-4-9(8)10(11)7-13/h1-7H
Formel vægt 174.048