missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylsulfonyl)ethanol, 94%, Thermo Scientific™

Catalog Number p-7024915
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
513-12-2
C4H10O3S
138.181
MFCD00041259
LJNDRJPZYNGSKZ-UHFFFAOYSA-N
2-ethylsulfonyl ethanol, ethylsulfonylethanol, ethanol, 2-ethylsulfonyl, ethylsulfonylethyl alcohol, ethyl 2-hydroxyethyl sulfone, unii-95yyw1049k, 2-ethanesulfonyl ethanol, ese, beta-ethylsulfonylethanol, 2-ethanesulfonyl-ethanol
10549
2-ethylsulfonylethanol
CCS(=O)(=O)CCO
This item is not returnable. View return policy
513-12-2
C4H10O3S
138.181
MFCD00041259
LJNDRJPZYNGSKZ-UHFFFAOYSA-N
2-ethylsulfonyl ethanol, ethylsulfonylethanol, ethanol, 2-ethylsulfonyl, ethylsulfonylethyl alcohol, ethyl 2-hydroxyethyl sulfone, unii-95yyw1049k, 2-ethanesulfonyl ethanol, ese, beta-ethylsulfonylethanol, 2-ethanesulfonyl-ethanol
10549
2-ethylsulfonylethanol
CCS(=O)(=O)CCO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 35°C to 41°C
Density 1.25
Boiling Point 153°C (2.5 mmHg)
Flash Point 188°C (370°F)
Quantity 5 g
Beilstein 1748754
Sensitivity Hygroscopic
Formula Weight 138.19
Percent Purity 94%
Chemical Name or Material 2-(Ethylsulfonyl)ethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'