Få mere at vide
(±)-2-Ethylhexyl-d17 Alcohol, CDN
Artikelnummer. 30324125
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324125 0.1 g 0.1g
30312742 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324125 Leverandør CDN Leverandørnr. CDND52470.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Ethyl-1-hexanol-d17
CAS 202480-75-9
Molekylær formel C8 2H17 H O
Mængde 0.1 g
InChI formel InChI=1 S/C8H18O/c1-3-5-6-8(4-2)7-9/h8-9 H,3-7H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8 D
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])O)C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,2,3,3,4,4,5,5,6,6,6-dodecadeuterio-2-(1,1,2,2,2-pentadeuterioethyl)hexan-1-ol
Molekylvægt (g/mol) 147.33
Formel vægt 147.2425 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity