missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Ethylbenzofuran, TRC

Product Code. 30388294
Change view
Click to view available options
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30388294 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388294 Supplier Toronto Research Chemicals Supplier No. TRCE8997705G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Ethylbenzofuran
CAS 3131-63-3
Molekylær formel C10H10O
Mængde 5 g
Synonym 2-Ethylbenzo[b]furan, 2-Ethylbenzofuran
InChI formel InChI=1S/C10H10O/c1-2-9-7-8-5-3-4-6-10(8)11-9/h3-7H,2H2,1H3
SMIL CCc1oc2ccccc2c1
Molekylvægt (g/mol) 146.19
Formel vægt 146.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.