Learn More
2-Ethylanthraquinone, Sigma-Aldrich™
Product Code. 15874641
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity Unit Size
15874641 100 g 100g
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15874641 Supplier Sigma Aldrich Supplier No. 8219340100
 Restricted Item
Forventet forsendelse: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 84-51-5
Melting Point 108.8°C
Density 1.27 g/cm3 at 21°C
MDL Number MFCD00001237
Quantity 100 g
Synonym 2-Ethylanthraquinone, 2-Ethylanthraquinone
Molecular Weight (g/mol) 236.27
Grade Research