Få mere at vide
2-[(Ethylamino)methyl]-4-nitrophenol-d5, TRC
Artikelnummer. 30379226
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30379226 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379226 Leverandør Toronto Research Chemicals Leverandørnr. TRCE898827100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[(Ethylamino)methyl]-4-nitrophenol-d5
CAS 1185157-36-1
Molekylær formel C9H7D5N2O3
Mængde 100 mg
InChI formel InChI=1S/C9H12N2O3/c1-2-10-6-7-5-8(11(13)14)3-4-9(7)12/h3-5,10,12H,2,6H2,1H3/i1D3,2D2
SMIL OC1=C(CNC([2H])([2H])C([2H])([2H])[2H])C=C([N+]([O-])=O)C=C1
Molekylvægt (g/mol) 201.23