missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylamino)ethanol, 98%, Thermo Scientific Chemicals

Catalog Number p-7022820
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
110-73-6
C4H11NO
89.138
MFCD00002841
MIJDSYMOBYNHOT-UHFFFAOYSA-N
2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
8072
2-(ethylamino)ethanol
CCNCCO
This item is not returnable. View return policy
110-73-6
C4H11NO
89.138
MFCD00002841
MIJDSYMOBYNHOT-UHFFFAOYSA-N
2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
8072
2-(ethylamino)ethanol
CCNCCO

2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.

Solubility
Miscible in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY
Melting Point -9°C
Boiling Point 168°C to 170°C
Flash Point 71°C (160°F)
Linear Formula CH3CH2NHCH2CH2OH
Quantity 100 mL
UN Number UN2922
Beilstein 635671
Formula Weight 89.14
Percent Purity 98%
Chemical Name or Material 2-(Ethylamino)ethanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'