missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Ethyl-d5-naphthalene, CDN

Artikelnummer. 30323980
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323980 0.5 g 0.5g
30312720 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323980 Leverandør CDN Leverandørnr. CDND71990.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Ethylnaphthalene-D5 (ethyl-D5)
CAS 1219805-14-7
Molekylær formel C12 D5 H7
Mængde 0.5 g
Synonym Naphthalene, 2-(ethyl-1,1,2,2,2-d5)- (ACI), 2-(Ethyl-1,1,2,2,2-d5)naphthalene (ACI), 2-Ethyl-d5-naphthalene, 2-Ethylnaphthalene-d5, 2-Ethylnaphthalene-D5 (ethyl-D5)
InChI formel InChI=1 S/C12H12/c1-2-10-7-8-11-5-3-4-6-12(11)9-10/h3-9 H,2H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])c1ccc2ccccc2c1
IUPAC navn 2-(1,1,2,2,2-pentadeuterioethyl)naphthalene
Molekylvægt (g/mol) 161.2545
Formel vægt 161.1253 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.