Få mere at vide
2-Ethyl-6-methylpyrazine, TRC
Artikelnummer. 30132913
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132913 100 mg
30133841 50 mg
17913375 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132913 Leverandør Toronto Research Chemicals Leverandørnr. TRCB438208100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13925-03-6
Molekylær formel C7H10N2
Molekylvægt (g/mol) 122.17
Synonym Pyrazine, 2-ethyl-6-methyl-, 2-Ethyl-6-methylpyrazine, 2-Methyl-6-ethylpyrazine, 6-Methyl-2-ethylpyrazine
IUPAC navn 2-ethyl-6-methylpyrazine
SMIL CCC1=NC(=CN=C1)C
Tekniske data
Kemisk navn eller materiale 2-Ethyl-6-methylpyrazine
Mængde 100 mg
InChI formel InChI=1S/C7H10N2/c1-3-7-5-8-4-6(2)9-7/h4-5H,3H2,1-2H3
Formel vægt 122.0844