Få mere at vide
2-Ethyl-6-methylaniline-d13, CDN
Artikelnummer. 30322275
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322275 0.25 g -
30320818 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322275 Leverandør CDN Leverandørnr. CDND59190.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Ethyl-6-methylaniline-d13
CAS 1219794-93-0
Molekylær formel C9 D13 N
Mængde 0.25 g
Synonym 2-(Ethyl-d5)-6-(methyl-d3)benzen-3,4,5-d3-amine-d2, 2-Ethyl-6-methylaniline-d13, 2-Ethyl-6-methylaniline-D13
InChI formel InChI=1 S/C9H13N/c1-3-8-6-4-5-7(2)9(8)10/h4-6 H,3,10H2,1-2H3/i1D3,2D3,3D2,4 D,5 D,6 D/hD2
SMIL [2 H]N([2 H])c1c(c([2 H])c([2 H])c([2 H])c1C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn N,N,3,4,5-pentadeuterio-2-(1,1,2,2,2-pentadeuterioethyl)-6-(trideuteriomethyl)aniline
Molekylvægt (g/mol) 148.2863
Formel vægt 148.1864 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity