Få mere at vide
2-Ethyl-2-phenylpropane-diamide Hydrate (Ethylphenylmalonamide Hydrate), Mikromol™
Artikelnummer. 30052367
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052367 100 mg
30052283 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052367 Leverandør Mikromol Leverandørnr. MM1030.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Ethyl-2-phenylpropane-diamide Hydrate (Ethylphenylmalonamide Hydrate)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Ethyl-2-phenylpropane-diamide Hydrate (Ethylphenylmalonamide Hydrate)
CAS 80866-90-6
Analyt- eller komponentnavne 2-Ethyl-2-phenylpropane-diamide Hydrate (Ethylphenylmalonamide Hydrate)
Forsendelsestilstand Room Temperature
Molekylær formel C11 H14 N2 O2 . H2 O
Certificeringer/compliance ISO 17025
Synonym Primidone Imp. A (EP), Primidone USP Related Compound A, Primidone USP RC A, 2-Ethyl-2-phenylpropane-diamide Hydrate, Primidone Imp. A (EP) as Hydrate, Ethylphenylmalonamide Hydrate, Primidone Related Compound A as Monohydrate, Primidone Impurity A as Hydrate
InChI formel InChI=1S/C11H14N2O2.H2O/c1-2-11(9(12)14,10(13)15)8-6-4-3-5-7-8;/h3-7H,2H2,1H3,(H2,12,14)(H2,13,15);1H2
SMIL O.CCC(C(=O)N)(C(=O)N)c1ccccc1
IUPAC navn 2-ethyl-2-phenylpropanediamide;hydrate
Molekylvægt (g/mol) 224.26
Mængde 100 mg
Formel vægt 224.1161 g/mol