Få mere at vide
2-Ethyl-2-methyl-succinonitrile-d3, TRC
Artikelnummer. 30375618
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30375618 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375618 Leverandør Toronto Research Chemicals Leverandørnr. TRCE92517210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Ethyl-2-methyl-succinonitrile-d3
CAS 1246817-05-9
Molekylær formel C7H7D3N2
Mængde 10 mg
Synonym 2-Ethyl-2-methyl-butanedinitrile-d3
InChI formel InChI=1S/C7H10N2/c1-3-7(2,6-9)4-5-8/h3-4H2,1-2H3/i1D3
SMIL [2H]C([2H])([2H])CC(C)(CC#N)C#N
IUPAC navn 2-methyl-2-(2,2,2-trideuterioethyl)butanedinitrile
Molekylvægt (g/mol) 125.19
Formel vægt 125.1032