Learn More
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, Sigma-Aldrich™
Product Code. 15829630
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Unit Size:
100g
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829630 2.5 kg 2.5kg
15809630 100 g 100g
15819630 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15829630 Supplier Sigma Aldrich Supplier No. 8083942500
 Restricted Item
Forventet forsendelse: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 77-99-6
Grade Research
Molecular Formula CH3CH2C(CH2OH)3
Molecular Weight (g/mol) 134.17
Specifications
Melting Point 58°C
MDL Number MFCD00004694
Quantity 2.5 kg
Synonym 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, Propylidynetris(methanol), 1,1,1-Trimethylolpropane, 1,1,1-Tris(hydroxymethyl)propane, 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol