Få mere at vide
2-Ethyl-2-hexenol (>85%), TRC
Artikelnummer. 30137604
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137604 250 mg
30138546 100 mg
30006781 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137604 Leverandør Toronto Research Chemicals Leverandørnr. TRCE817893250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 50639-00-4
Molekylær formel C8 H16 O
Molekylvægt (g/mol) 128.21
Synonym 2-Ethyl-2-hexenol (>85%), 2-Ethyl-2-hexen-1-ol
IUPAC navn (Z)-2-ethylhex-2-en-1-ol
SMIL CCC\C=C(\CC)/CO
Tekniske data
Kemisk navn eller materiale 2-Ethyl-2-hexenol
Mængde 250 mg
InChI formel InChI=1S/C8H16O/c1-3-5-6-8(4-2)7-9/h6,9H,3-5,7H2,1-2H3/b8-6-
Formel vægt 128.1201