Få mere at vide
2-Ethyl-1-hexanol-d17, TRC
Artikelnummer. 30379052
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30379052 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379052 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91875750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Ethyl-1-hexanol-d17
CAS 202480-75-9
Molekylær formel C8 2H17 H O
Mængde 50 mg
Synonym 1-Hexan-1,1,2,3,3,4,4,5,5,6,6,6-d12-ol, 2-(ethyl-d5)- (9CI, ACI), 2-(Ethyl-d5)-1-hexan-1,1,2,3,3,4,4,5,5,6,6,6-d12-ol (ACI), (±)-2-Ethylhexyl-d17 Alcohol, 2-Ethyl-1-hexanol-d17, 2-Ethyl-1-hexanol-D17
InChI formel InChI=1S/C8H18O/c1-3-5-6-8(4-2)7-9/h8-9H,3-7H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(C([2H])([2H])O)C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,1,2,3,3,4,4,5,5,6,6,6-dodecadeuterio-2-(1,1,2,2,2-pentadeuterioethyl)hexan-1-ol
Molekylvægt (g/mol) 147.33
Formel vægt 147.2425