missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7030618
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
269409-99-6
C15H21BO4
276.139
MFCD03453660
RFBZWPFBCXBBJS-UHFFFAOYSA-N
ethyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid pinacol ester, ethyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid, pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester, ethyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-carboethoxyphenylboronic acid pinacol ester, 2-ethoxycarbonyl benzeneboronic acid pinacol ester, ethyl 2-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzoate
2773962
ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C(=O)OCC
This item is not returnable. View return policy
269409-99-6
C15H21BO4
276.139
MFCD03453660
RFBZWPFBCXBBJS-UHFFFAOYSA-N
ethyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid pinacol ester, ethyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid, pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester, ethyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-carboethoxyphenylboronic acid pinacol ester, 2-ethoxycarbonyl benzeneboronic acid pinacol ester, ethyl 2-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzoate
2773962
ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 57°C to 62°C
Quantity 1 g
Formula Weight 276.14
Percent Purity 97%
Chemical Name or Material 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'