Få mere at vide
2-Ethoxy-1-[[2'-[(hydroxyamino)iminomethyl][1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic Acid Methyl Ester, TRC
Artikelnummer. 30180247
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180247 100 mg
30180428 50 mg
30180265 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180247 Leverandør Toronto Research Chemicals Leverandørnr. TRCE892030100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 147403-65-4
Molekylær formel C25H24N4O4
Molekylvægt (g/mol) 444.48
Synonym Methyl 2-Ethoxy-1-[[2'-(hydroxyamidino)biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate
SMIL CCOc1nc2cccc(C(=O)OC)c2n1Cc3ccc(cc3)c4ccccc4C(=N)NO
Tekniske data
Kemisk navn eller materiale 2-Ethoxy-1-((2'-((hydroxyamino)iminomethyl)(1,1'-biphenyl)-4-yl)methyl)-1H-benzimidazole-7-carboxylic Acid Methyl Ester
Mængde 100 mg
InChI formel InChI=1S/C25H24N4O4/c1-3-33-25-27-21-10-6-9-20(24(30)32-2)22(21)29(25)15-16-11-13-17(14-12-16)18-7-4-5-8-19(18)23(26)28-31/h4-14,31H,3,15H2,1-2H3,(H2,26,28)
Formel vægt 444.1798