missing translation for 'onlineSavingsMsg'
Learn More

2-Diphenylphosphino-2'-methylbiphenyl, 98%, Thermo Scientific™

Catalog Number 15478258
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
402822-72-4
C25H21P
352.42
MFCD19441232
GUENDLLDUNRPIC-UHFFFAOYSA-N
2'-methyl-1,1'-biphenyl-2-yl diphenylphosphine, 2-diphenylphosphino-2'-methylbiphenyl, phosphine, 2'-methyl 1,1'-biphenyl-2-yl diphenyl, 2'-methyl-1,1'-biphenyl-2-yl diphenylphosphane, 2-diphenylphosphino-2-methylbiphenyl, 2'-methyl-2-biphenylyldiphenylphosphine, 2-2-methylphenyl phenyl diphenylphosphane, 2'-methyl 1,1'-biphenyl-2-yl diphenyl phosphane
15531745
[2-(2-methylphenyl)phenyl]-diphenylphosphane
CC1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
402822-72-4
C25H21P
352.42
MFCD19441232
GUENDLLDUNRPIC-UHFFFAOYSA-N
2'-methyl-1,1'-biphenyl-2-yl diphenylphosphine, 2-diphenylphosphino-2'-methylbiphenyl, phosphine, 2'-methyl 1,1'-biphenyl-2-yl diphenyl, 2'-methyl-1,1'-biphenyl-2-yl diphenylphosphane, 2-diphenylphosphino-2-methylbiphenyl, 2'-methyl-2-biphenylyldiphenylphosphine, 2-2-methylphenyl phenyl diphenylphosphane, 2'-methyl 1,1'-biphenyl-2-yl diphenyl phosphane
15531745
[2-(2-methylphenyl)phenyl]-diphenylphosphane
CC1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 352.41
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Diphenylphosphino-2'-methylbiphenyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'