Få mere at vide
2-Diisopropylaminoethyl Chloride Hydrochloride-d14, TRC
Artikelnummer. 30376798
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30376798 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376798 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4554242.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Diisopropylaminoethyl Chloride Hydrochloride-d14
CAS 75174-04-8
Molekylær formel C8H4D14ClN . HCl
Mængde 2.5 mg
Synonym (beta-Chloroethyl)diisopropylamine Hydrochloride-d14, 2-(Diisopropylamino)ethyl Chloride Hydrochloride-d14, 2-(N,N-Diisopropylamino)ethyl Chloride Hydrochloride-d14, 2-Chloro-N,N-bis(isopropyl)ethanamine Hydrochloride-d14, 2-Chloro-N,N-diisopropylethylamine Hydrochloride-d14, 2-Chloroethyldiisopropylamine Hydrochloride-d14, N,N-Diisopropyl-2-Chloroethylamine Hydrochloride-d14, N-(2-Chloroethyl)-N-isopropylpropan-2-amine Hydrochloride-d14, N-(2-Chloroethyl)diisopropylamine Hydrochloride-d14
InChI formel InChI=1S/C8H18ClN.ClH/c1-7(2)10(6-5-9)8(3)4;/h7-8H,5-6H2,1-4H3;1H/i1D3,2D3,3D3,4D3,7D,8D;
SMIL Cl.[2H]C([2H])([2H])C([2H])(N(CCCl)C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn N-(2-chloroethyl)-1,1,1,2,3,3,3-heptadeuterio-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)propan-2-amine;hydrochloride
Molekylvægt (g/mol) 214.23
Formel vægt 213.1773