missing translation for 'onlineSavingsMsg'
Learn More

2-Diethylaminoethyl 4-aminobenzoate hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-7023682
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
51-05-8
C13H21ClN2O2
272.77
MFCD00013000
HCBIBCJNVBAKAB-UHFFFAOYSA-N
procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor
5795
CHEBI:8431
[H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1
This item is not returnable. View return policy
51-05-8
C13H21ClN2O2
272.77
MFCD00013000
HCBIBCJNVBAKAB-UHFFFAOYSA-N
procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor
5795
CHEBI:8431
[H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1

A sodium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 155°C to 157°C
Boiling Point 195°C to 196°C (17 mmHg)
Odor Odorless
Quantity 50 g
UN Number UN2811
Sensitivity Light sensitive
Merck Index 14,7757
IUPAC Name 2-(diethylamino)ethyl 4-aminobenzoate;hydrochloride
Formula Weight 272.78
Percent Purity 99%
Chemical Name or Material 2-Diethylaminoethyl 4-aminobenzoate hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'