Få mere at vide
2-Desmethyl 4-Methyl Prilocaine, TRC
Artikelnummer. 30364239
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364239 50 mg
30352494 100 mg
30359410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30364239 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29228550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 744961-76-0
Molekylær formel C13 H20 N2 O
Molekylvægt (g/mol) 220.31
Synonym 2-(Propylamino)-p-propionotoluidide, (RS)-N-(4-Methylphenyl)-2-(propylamino)propanamide
IUPAC navn N-(4-methylphenyl)-2-(propylamino)propanamide
SMIL CCCNC(C)C(=O)Nc1ccc(C)cc1
Tekniske data
Kemisk navn eller materiale (RS)-N-(4-methylphenyl)-2-(propylamino)propanamide
Mængde 50 mg
InChI formel InChI=1S/C13H20N2O/c1-4-9-14-11(3)13(16)15-12-7-5-10(2)6-8-12/h5-8,11,14H,4,9H2,1-3H3,(H,15,16)
Formel vægt 220.1576