missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Desfluoro Fluconazole, TRC

Artikelnummer. 30363929
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363929 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30363929 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22885550MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-(4-Fluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
CAS 81886-51-3
Molekylær formel C13 H13 F N6 O
Mængde 50 mg
Synonym 2-(4-Fluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol, Fluconazole Imp. D (EP)
InChI formel InChI=1S/C13H13FN6O/c14-12-3-1-11(2-4-12)13(21,5-19-9-15-7-17-19)6-20-10-16-8-18-20/h1-4,7-10,21H,5-6H2
SMIL OC(Cn1cncn1)(Cn2cncn2)c3ccc(F)cc3
IUPAC navn 2-(4-fluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol
Molekylvægt (g/mol) 288.28
Formel vægt 288.1135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.