Få mere at vide
2-Descarboxy-2-cyano Telmisartan, TRC
Artikelnummer. 30356121
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356121 100 mg
30348871 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30356121 Leverandør Toronto Research Chemicals Leverandørnr. TRCD289075100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Descarboxy-2-cyano Telmisartan
CAS 144702-27-2
Molekylær formel C33 H29 N5
Mængde 100 mg
Synonym 4'-[[4-Methyl-6-(1-methyl-1H-benz-imidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]-methyl]biphenyl-2-carbonitrile, 2-Descarboxy-2-cyano Telmisartan, 4'-[(1,4'-Dimethyl-2'-propyl[2,6'-bi-1H-benzimidazol]-1'-yl)methyl][1,1'-biphenyl]-2-carbonitrile, Telmisartan Imp G (EP)
InChI formel InChI=1S/C33H29N5/c1-4-9-31-36-32-22(2)18-26(33-35-28-12-7-8-13-29(28)37(33)3)19-30(32)38(31)21-23-14-16-24(17-15-23)27-11-6-5-10-25(27)20-34/h5-8,10-19H,4,9,21H2,1-3H3
SMIL CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)c4ccccc4C#N)c5nc6ccccc6n5C
IUPAC navn 2-[4-[[4-methyl-6-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzonitrile
Molekylvægt (g/mol) 495.62
Formel vægt 495.2423