Få mere at vide
2'-Deoxyuridine-5,6-d2, CDN
Artikelnummer. 30324766
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324766 0.01 g
30320748 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324766 Leverandør CDN Leverandørnr. CDND53520.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2'-Deoxyuridine-5,6-D2
CAS 40632-23-3
Molekylær formel C9 D2 H10 N2 O5
Mængde 0.01 g
Synonym Uridine-5,6-d2, 2'-deoxy- (9 CI), 2'-Deoxy-5,6-dideuteriouridine, 2'-Deoxyuridine-5,6-d2, 2'-Deoxyuridine-5,6-D2, 1-(2-Deoxy-β-D-ribofuranosyl)uracil-5,6-d2, 1-(2-Deoxy-β-D-ribofuranosyl)-2,4(1 H,3 H)-pyrimidinedione-5,6-d2, 1-(2-Deoxy-β-D-ribofuranosyl)-2,4(1 H,3 H)-pyrimidinedione-5,6-d2
InChI formel InChI=1 S/C9H12N2O5/c12-4-6-5(13)3-8(16-6)11-2-1-7(14)10-9(11)15/h1-2,5-6,8,12-13 H,3-4H2,(H,10,14,15)/t5-,6+,8+/m0/s1/i1D,2 D
SMIL [2 H]C1=C([2 H])C(=O)NC(=O)N1[C@H]2 C[C@H](O)[C@@H](CO)O2
IUPAC navn 5,6-dideuterio-1-[(2 R,4 S,5 R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Molekylvægt (g/mol) 230.2143
Formel vægt 230.0872 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity