missing translation for 'onlineSavingsMsg'
Få mere at vide

2'-Deoxyguanosine-5'-diphosphate Disodium Salt, TRC

Artikelnummer. 30368358
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368358 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30368358 Leverandør Toronto Research Chemicals Leverandørnr. TRCD24244350MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2'-Deoxyguanosine-5'-diphosphate Disodium Salt
CAS 78101-74-3
Molekylær formel C10 H13 N5 Na2 O10 P2
Mængde 50 mg
InChI formel InChI=1S/C10H15N5O10P2.2Na/c11-10-13-8-7(9(17)14-10)12-3-15(8)6-1-4(16)5(24-6)2-23-27(21,22)25-26(18,19)20;;/h3-6,16H,1-2H2,(H,21,22)(H2,18,19,20)(H3,11,13,14,17);;/q;2*+1/p-2/t4-,5+,6+;;/m0../s1
SMIL NC1=NC(=O)c2ncn([C@H]3C[C@H](O)[C@@H](COP(=O)(O[Na])OP(=O)(O)O[Na])O3)c2N1
IUPAC navn [[(2R,3S,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy-[hydroxy(sodiooxy)phosphoryl]oxyphosphoryl]oxysodium
Molekylvægt (g/mol) 471.165
Formel vægt 470.993
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.