missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Deoxycytidine-5'-triphosphate disodium salt, Thermo Scientific™
Click to view available options
Quantity:
25 mg
250 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2′-Deoxycytidine 5′-triphosphate disodium salt is a nucleoside triphosphatase used for DNA synthesis with DNA polymerases and reverse transcriptases. 2′-Deoxycytidine 5′-triphosphate disodium salt is one of the nucleoside triphosphates used for DNA polymerase driven reactions such as polymerase chain reaction, DNA sequencing and other molecular biology methods.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
2′-Deoxycytidine 5′-triphosphate disodium salt is a nucleoside triphosphatase used for DNA synthesis with DNA polymerases and reverse transcriptases. 2′-Deoxycytidine 5′-triphosphate disodium salt is one of the nucleoside triphosphates used for DNA polymerase driven reactions such as polymerase chain reaction, DNA sequencing and other molecular biology methods.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Specifications
Color | White |
Quantity | 25 mg |
Formula Weight | 511.12 |
Physical Form | Powder |
Chemical Name or Material | 2'-Deoxycytidine-5'-triphosphate disodium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'