Få mere at vide
2’-Deoxy Cytidine-5,6-d2, TRC
Artikelnummer. 30361423
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30361423 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361423 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2326185MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2'-Deoxycytidine-5,6-D2
CAS 1186526-91-9
Molekylær formel C9 D2 H11 N3 O4
Mængde 5 mg
Synonym Cytidine-5,6-d2, 2'-deoxy-, 2'-Deoxycytidine-5,6-D2, 5,6-[2H] 2'-Deoxycytidine
InChI formel InChI=1S/C9H13N3O4/c10-7-1-2-12(9(15)11-7)8-3-5(14)6(4-13)16-8/h1-2,5-6,8,13-14H,3-4H2,(H2,10,11,15)/t5-,6+,8+/m0/s1/i1D,2D
SMIL [2H]C1=C([2H])C(=NC(=O)N1[C@H]2C[C@H](O)[C@@H](CO)O2)N
IUPAC navn 4-amino-5,6-dideuterio-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
Molekylvægt (g/mol) 229.2295
Formel vægt 229.1032