Få mere at vide
2-Demethoxy-4-methoxy Urapidil, TRC
Artikelnummer. 30133504
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133504 10 mg
30133258 50 mg
30009249 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133504 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23113010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34661-79-5
Molekylær formel C20H29N5O3
Molekylvægt (g/mol) 387.48
Synonym 6-[[3-[4-(4-Methoxyphenyl)-1-piperazinyl]propyl]amino]-1,3-dimethyl-2,4(1H,3H)-pyrimidinedione;
SMIL COc1ccc(cc1)N2CCN(CCCNC3=CC(=O)N(C)C(=O)N3C)CC2
Tekniske data
Kemisk navn eller materiale 2-Demethoxy-4-methoxy Urapidil
Mængde 10 mg
InChI formel InChI=1S/C20H29N5O3/c1-22-18(15-19(26)23(2)20(22)27)21-9-4-10-24-11-13-25(14-12-24)16-5-7-17(28-3)8-6-16/h5-8,15,21H,4,9-14H2,1-3H3
Formel vægt 387.227