Få mere at vide
2-(Cyclopropylmethoxy-d4)-acetic Acid, TRC
Artikelnummer. 30350983
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30350983 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350983 Leverandør Toronto Research Chemicals Leverandørnr. TRCC988687100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(Cyclopropylmethoxy-d4)-acetic Acid
CAS 1346602-05-8
Molekylær formel C6H6D4O3
Mængde 100 mg
Synonym (Cyclopropylmethoxy-d4)acetic Acid
InChI formel InChI=1S/C6H10O3/c7-6(8)4-9-3-5-1-2-5/h5H,1-4H2,(H,7,8)/i1D2,2D2
SMIL [2H]C1([2H])C(COCC(=O)O)C1([2H])[2H]
IUPAC navn 2-[(2,2,3,3-tetradeuteriocyclopropyl)methoxy]acetic acid
Molekylvægt (g/mol) 134.17
Formel vægt 134.0881