Få mere at vide
2-Cyclopropylethyl-1,1,2,2-d4-amine, CDN
Artikelnummer. 30324841
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324841 0.1 g 0.1g
30315042 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324841 Leverandør CDN Leverandørnr. CDND70010.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Cyclopropylethylamine-1,1,2,2-D4
CAS 1219795-00-2
Molekylær formel C5 D4 H7 N
Mængde 0.1 g
Synonym 2-Cyclopropylethyl-1,1,2,2-d4-amine, 2-Cyclopropylethan-d4-amine, 2-Cyclopropylethylamine-1,1,2,2-D4
InChI formel InChI=1 S/C5H11N/c6-4-3-5-1-2-5/h5H,1-4,6H2/i3D2,4D2
SMIL [2 H]C([2 H])(N)C([2 H])([2 H])C1CC1
IUPAC navn 2-cyclopropyl-1,1,2,2-tetradeuterioethanamine
Molekylvægt (g/mol) 89.1722
Formel vægt 89.1143 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity