Få mere at vide
2-Cyclopentyl-2-hydroxy-2-phenylacetic Acid, TRC
Artikelnummer. 30003415
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003415 5 g
17984125 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30003415 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9893155G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 427-49-6
Molekylær formel C13H16O3
Molekylvægt (g/mol) 220.26
Synonym 2-(RS)-Cyclopentyl-2-hydroxy-2-phenylacetic Acid, Glycopyrronium Bromide Imp. J (EP), alpha-Cyclopentylmandelic Acid, Benzeneacetic acid, α-cyclopentyl- α-hydroxy- (9CI), Mandelic acid, α-cyclopentyl- (6CI,7CI,8CI), α-Cyclopentylmandelic acid, Cyclopentylphenylglycolic acid, NSC 96637
IUPAC navn 2-cyclopentyl-2-hydroxy-2-phenylacetic acid
SMIL OC(=O)C(O)(C1CCCC1)c2ccccc2
Tekniske data
Kemisk navn eller materiale 2-(RS)-Cyclopentyl-2-hydroxy-2-phenylacetic Acid
Renhedsgrad noter HPLC
Mængde 5 g
InChI formel InChI=1S/C13H16O3/c14-12(15)13(16,11-8-4-5-9-11)10-6-2-1-3-7-10/h1-3,6-7,11,16H,4-5,8-9H2,(H,14,15)
Formel vægt 220.1099
Procent renhed >95%