Få mere at vide
2-Cyclohexylidene-cyclohexanone, TRC
Artikelnummer. 30367244
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367244 50 mg
30368539 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367244 Leverandør Toronto Research Chemicals Leverandørnr. TRCC99570050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1011-12-7
Molekylær formel C12H18O
Molekylvægt (g/mol) 178.27
Synonym 2-Cyclohexylidenecyclohexanone, Dianon, Bicyclohexyliden-2-one
IUPAC navn 2-cyclohexylidenecyclohexan-1-one
SMIL O=C1CCCCC1=C2CCCCC2
Tekniske data
Kemisk navn eller materiale 2-Cyclohexylidene-cyclohexanone
Mængde 50 mg
InChI formel InChI=1S/C12H18O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H2
Formel vægt 178.1358