Learn More
2-Cyanothiazole, 95%, Thermo Scientific Chemicals
Catalog Number p-7054965
Click to view available options
Quantity:
250 mg
1 g
5 g
1452-16-0
C4H2N2S
110.134
MFCD09702472
ZIRGWUZHKJDYKT-UHFFFAOYSA-N
2-cyanothiazole, thiazole-2-carbonitrile, 2-thiazolecarbonitrile, 2-cyanothiazol
15111480
1,3-thiazole-2-carbonitrile
C1=CSC(=N1)C#N
1452-16-0
C4H2N2S
110.134
MFCD09702472
ZIRGWUZHKJDYKT-UHFFFAOYSA-N
2-cyanothiazole, thiazole-2-carbonitrile, 2-thiazolecarbonitrile, 2-cyanothiazol
15111480
1,3-thiazole-2-carbonitrile
C1=CSC(=N1)C#N
2-Cyanothiazole is used as a intermediate for pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Cyanothiazole is used as a intermediate for pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3439
Solubility Information Slightly soluble in water.
Formula Weight 110.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Cyanothiazole