Få mere at vide
2-Cyanopyridine-d4, CDN
Artikelnummer. 30319756
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319756 0.25 g
30317735 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319756 Leverandør CDN Leverandørnr. CDND63760.25G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Cyanopyridine-D4
CAS 1219795-17-1
Molekylær formel C6 D4 N2
Mængde 0.25 g
Synonym 2-Pyridine-3,4,5,6-d4-carbonitrile (ACI), 2-Cyanopyridine-3,4,5,6-d4, 2-Cyanopyridine-d4, 2-Cyanopyridine-D4
InChI formel InChI=1 S/C6H4N2/c7-5-6-3-1-2-4-8-6/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1nc(C#N)c([2 H])c([2 H])c1[2 H]
IUPAC navn 3,4,5,6-tetradeuteriopyridine-2-carbonitrile
Molekylvægt (g/mol) 108.134
Formel vægt 108.0626 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity