Få mere at vide
2-Cyanoacetohydrazide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Cyanoacetohydrazide |
| CAS | 140-87-4 |
| Molekylær formel | C3H5N3O |
| Mængde | 10 g |
| Synonym | Cyano-acetic Acid Hydrazide, (Cyanoacetyl)hydrazine, (alpha-Cyanoacetyl)hydrazine, 2-Cyanoacethydrazide, 2-Cyanoacetohydrazide, 2-Cyanoacetylhydrazide, AB-42, Armazal, Cianazil, Cyacetacid, Cyacetacide, Cyacetazid, Cyacetazide, Cyanacethydrazide, Cyanacetic Acid Hydrazide, Cyanacetohydrazide, Cyanacetylhydrazide, Cyanazide, Cyanizide, Cyanoacethydrazide, Cyanoacetic Acid Hydrazide, Cyanoacetic Hydrazide, Cyanoacetohydrazide, Cyanoacetyl Hydrazide, Cyanoethanoic Hydrazide, Cyazid, Cyazide, Dictycide, Dictyzide, Helmox, Hidacian, Hidaciann, Leandin, Mackreazid, Malonitrile Hydrazide, Malononitrile Hydrazide, NSC 24261, Neohydrazid, Reacid, Reazid, Reazide, Tsiazid, alpha-Cyanoacetohydrazide |
| InChI formel | InChI=1S/C3H5N3O/c4-2-1-3(7)6-5/h1,5H2,(H,6,7) |
| SMIL | NNC(=O)CC#N |
| IUPAC navn | 2-cyanoacetohydrazide |
| Molekylvægt (g/mol) | 99.09 |
| Formel vægt | 99.0433 |