missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3,5-difluoropyridine, 98%, Thermo Scientific Chemicals

Catalog Number 11345565
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
298709-29-2
C6H2F2N2
140.093
MFCD03788759
WLBIFECTHKFYKV-UHFFFAOYSA-N
2-cyano-3,5-difluoropyridine, 3,5-difluoropicolinonitrile, 3,5-difluoro-2-cyanopyridine, 2-pyridinecarbonitrile, 3,5-difluoro, 3,5-difluoropicolinitrile, abbypharma ap-15-5175, 2-cyano-3,5-difluorpyridine, 3,5-difluoro-2-pyridinecarbonitrile, pubchem15234, acmc-209z3h
2783253
3,5-difluoropyridine-2-carbonitrile
C1=C(C=NC(=C1F)C#N)F
This item is not returnable. View return policy
298709-29-2
C6H2F2N2
140.093
MFCD03788759
WLBIFECTHKFYKV-UHFFFAOYSA-N
2-cyano-3,5-difluoropyridine, 3,5-difluoropicolinonitrile, 3,5-difluoro-2-cyanopyridine, 2-pyridinecarbonitrile, 3,5-difluoro, 3,5-difluoropicolinitrile, abbypharma ap-15-5175, 2-cyano-3,5-difluorpyridine, 3,5-difluoro-2-pyridinecarbonitrile, pubchem15234, acmc-209z3h
2783253
3,5-difluoropyridine-2-carbonitrile
C1=C(C=NC(=C1F)C#N)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 30°C to 34°C
Quantity 1 g
Formula Weight 140.09
Percent Purity 98%
Chemical Name or Material 2-Cyano-3,5-difluoropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'