missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorotoluene, 98%, Thermo Scientific Chemicals

Catalog Number p-3757283
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
95-49-8
C7H7Cl
126.58
MFCD00000562
IBSQPLPBRSHTTG-UHFFFAOYSA-N
2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene
7238
1-chloro-2-methylbenzene
CC1=CC=CC=C1Cl
This item is not returnable. View return policy
95-49-8
C7H7Cl
126.58
MFCD00000562
IBSQPLPBRSHTTG-UHFFFAOYSA-N
2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene
7238
1-chloro-2-methylbenzene
CC1=CC=CC=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -36.0°C
CAS Min % 97.5
CAS Max % 100.0
Density 1.0800g/mL
Boiling Point 157.0°C to 159.0°C
Quantity 5 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5240 to 1.5260
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.08
Linear Formula CH3C6H4Cl
Beilstein 05, 290
Merck Index 15, 2175
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol, benzene, chlroform and, ether
Formula Weight 126.59
Percent Purity 98%
Chemical Name or Material 2-Chlorotoluene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chlorotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H361d-Suspected of damaging the unborn child.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'