missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorotoluene, 98%, ACROS Organics™

Brand:  Acros Organics 150205000

Additional Details : CAS Number : 95-49-8 Weight : 0.59000kg Transport : UN number : 2238 Chem class : 3 Pack group : III


Packaging Glass bottle
Quantity 500mL

 View more versions of this product

Product Code. 10477920

  • POA / 500mL

Currently not in stock

Please to purchase this item. Need a web account? Register with us today!

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on +45 70 27 99 20 for assistance.
View alternative products »


Description

Description

Specifications

Chemical Identifiers

95-49-8
126.59
IBSQPLPBRSHTTG-UHFFFAOYSA-N
7238
CC1=CC=CC=C1Cl
C7H7Cl
MFCD00000562
2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene
1-chloro-2-methylbenzene

Specifications

2-Chlorotoluene
49°C
Glass bottle
1.08
157.0°C to 159.0°C
-36.0°C
95-49-8
100.0
C7H7Cl
MFCD00000562
15, 2175
Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol, benzene, chlroform and, ether
CC1=CC=CC=C1Cl
126.59
126.59
98%
1.0800g/mL
Authentic
1.5240 to 1.5260
0.015% max.
Undesignated
500mL
97.5
97.5% min. (GC)
CH3C6H4Cl
05, 290
2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene
IBSQPLPBRSHTTG-UHFFFAOYSA-N
1-chloro-2-methylbenzene
7238
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Chlorotoluene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H361d-Suspected of damaging the unborn child.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents