Learn More
2-chloropyrimidine-5-carboxylic acid, 97%, ACROS Organics™
Brand: Acros Organics 429470050
Additional Details : CAS Number : 374068-01-6 Weight : 0.05500kg
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
374068-01-6 | |
158.54 | |
2-chloro-pyrimidine-5-carboxylic acid, 2-chloro-5-carboxypyrimidine, 5-carboxy-2-chloropyrimidine, 5-pyrimidinecarboxylicacid, 2-chloro, 2-chloro-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylic acid, 2-chloro, 2-chloropyrimidine-5-carboxylicacid, acmc-1aig3, 2-chloropyrimidine 5-carboxylic acid, 2-chloro-5-pyrimidine carboxylic acid | |
2-chloropyrimidine-5-carboxylic acid |
C5H3ClN2O2 | |
DUCXUPKLVVSJKA-UHFFFAOYSA-N | |
11094906 | |
C1=C(C=NC(=N1)Cl)C(=O)O |
Specifications
2-Chloropyrimidine-5-carboxylic acid | |
Glass bottle | |
125.0°C to 127.5°C | |
374068-01-6 | |
100.0 | |
C5H3ClN2O2 | |
DUCXUPKLVVSJKA-UHFFFAOYSA-N | |
2-chloropyrimidine-5-carboxylic acid | |
11094906 | |
Powder or Crystalline Powder |
Authentic | |
White | |
5g | |
96.0 | |
96% min. (HPLC) | |
2-chloro-pyrimidine-5-carboxylic acid, 2-chloro-5-carboxypyrimidine, 5-carboxy-2-chloropyrimidine, 5-pyrimidinecarboxylicacid, 2-chloro, 2-chloro-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylic acid, 2-chloro, 2-chloropyrimidine-5-carboxylicacid, acmc-1aig3, 2-chloropyrimidine 5-carboxylic acid, 2-chloro-5-pyrimidine carboxylic acid | |
C1=C(C=NC(=N1)Cl)C(=O)O | |
158.54 | |
158.54 | |
97% |
Safety and Handling
- 2-Chloropyrimidine-5-carboxylic acid, 97%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.