missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrazine, 98%, Thermo Scientific Chemicals

Catalog Number p-2698191
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14508-49-7
C4H3ClN2
114.53
MFCD00006124
GELVZYOEQVJIRR-UHFFFAOYSA-N
chloropyrazine, pyrazine, chloro, pyrazine, 2-chloro, 2-chloro-pyrazine, 2-chloro pyrazine, chioropyrazine, chloropyrazin, chloro-pyrazine, 6-chloropyrazine, 2-chloropyrazine
73277
2-chloropyrazine
ClC1=CN=CC=N1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14508-49-7
C4H3ClN2
114.53
MFCD00006124
GELVZYOEQVJIRR-UHFFFAOYSA-N
chloropyrazine, pyrazine, chloro, pyrazine, 2-chloro, 2-chloro-pyrazine, 2-chloro pyrazine, chioropyrazine, chloropyrazin, chloro-pyrazine, 6-chloropyrazine, 2-chloropyrazine
73277
2-chloropyrazine
ClC1=CN=CC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2800g/mL
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5390
Quantity 10 g
Specific Gravity 1.28
Solubility Information Solubility in water: insoluble in water
Formula Weight 114.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloropyrazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloropyrazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'