Learn More
2-Chloropropane, Sigma-Aldrich™
Product Code. 15898840
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898840 100 mL 100mL
15806330 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15898840 Supplier Sigma Aldrich Supplier No. 8028090100
 Restricted Item
Forventet forsendelse: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-29-6
Grade Research
Molecular Formula (CH3)2CHCl
Molecular Weight (g/mol) 78.54
Specifications
Melting Point -118°C
Density 0.86 g/cm3 at 20°C
MDL Number MFCD00000867
Quantity 100 mL
Synonym 2-Chloropropane, Isopropyl chloride, 2-Chloropropane