Få mere at vide
(2-Chlorophenyl)diphenylmethane, TRC
Artikelnummer. 30359295
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359295 1 g
30358650 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30359295 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3778951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56153-60-7
Molekylær formel C19 H15 Cl
Molekylvægt (g/mol) 278.78
Synonym (2-Chlorophenyl)diphenylmethane, 1-Chloro-2-(diphenylmethyl)benzene
IUPAC navn 1-benzhydryl-2-chlorobenzene
SMIL Clc1ccccc1C(c2ccccc2)c3ccccc3
Tekniske data
Kemisk navn eller materiale (2-Chlorophenyl)diphenylmethane
Mængde 1 g
InChI formel InChI=1S/C19H15Cl/c20-18-14-8-7-13-17(18)19(15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14,19H
Formel vægt 278.0862