Få mere at vide
(2-Chlorophenyl)diphenylmethane, Mikromol™
Artikelnummer. 30049301
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049301 100 mg
30052461 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049301 Leverandør Mikromol Leverandørnr. MM0015.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2-Chlorophenyl)diphenylmethane
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2-Chlorophenyl)diphenylmethane
CAS 56153-60-7
Analyt- eller komponentnavne (2-Chlorophenyl)diphenylmethane
Forsendelsestilstand Room Temperature
Molekylær formel C19 H15 Cl
Certificeringer/compliance ISO 17025
Synonym (2-Chlorophenyl)diphenylmethane
InChI formel InChI=1S/C19H15Cl/c20-18-14-8-7-13-17(18)19(15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14,19H
SMIL Clc1ccccc1C(c2ccccc2)c3ccccc3
IUPAC navn 1-benzhydryl-2-chlorobenzene
Molekylvægt (g/mol) 278.78
Mængde 100 mg
Formel vægt 278.0862 g/mol