Få mere at vide
2-Chlorophenol-d4, TRC
Artikelnummer. 30362540
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30362540 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362540 Leverandør Toronto Research Chemicals Leverandørnr. TRCC993148100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chlorophenol-3,4,5,6-D4
CAS 93951-73-6
Renhedsgrad noter HPLC
Molekylær formel C6 2H4 H Cl O
Mængde 100 mg
Synonym Phen-2,3,4,5-d4-ol, 6-chloro- (9CI), 2-Chlorophenol D4, 2-Chlorophenol (ring-D4), 2-Chloro-3,4,5,6-tetradeuteriophenol
InChI formel InChI=1S/C6H5ClO/c7-5-3-1-2-4-6(5)8/h1-4,8H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c(Cl)c(O)c1[2H]
IUPAC navn 2-chloro-3,4,5,6-tetradeuteriophenol
Molekylvægt (g/mol) 132.58
Formel vægt 132.028
Procent renhed >95