Få mere at vide
2-Chlorophenol D4 (3,4,5,6 D4), Dr. Ehrenstorfer
Artikelnummer. 30324162
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30324162 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324162 Leverandør Dr. Ehrenstorfer Leverandørnr. DRECA11470100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chlorophenol-3,4,5,6-D4
CAS 93951-73-6
Molekylær formel C6 2H4 H Cl O
Mængde 25 mg
Synonym Phen-2,3,4,5-d4-ol, 6-chloro- (9CI), 2-Chlorophenol D4, 2-Chlorophenol (ring-D4), 2-Chloro-3,4,5,6-tetradeuteriophenol
InChI formel InChI=1S/C6H5ClO/c7-5-3-1-2-4-6(5)8/h1-4,8H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c(Cl)c(O)c1[2H]
IUPAC navn 2-chloro-3,4,5,6-tetradeuteriophenol
Molekylvægt (g/mol) 132.58
Formel vægt 132.028 g/mol