Få mere at vide
2-Chloronaphthalene-d7, TRC
Artikelnummer. 30361908
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30361908 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361908 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3737311MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Chloronaphthalene-d7
CAS 93951-84-9
Molekylær formel C10 2H7 Cl
Mængde 1 mg
Synonym Naphthalene-1,2,3,4,5,6,8-d7, 7-chloro- (9CI), 2-Chloronaphthalene D7, 2-Chloro-1,3,4,5,6,7,8-heptadeuterionaphthalene, 2-Chloronaphthalene-1,3,4,5,6,7,8-D7
InChI formel InChI=1S/C10H7Cl/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H/i1D,2D,3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c2c([2H])c(Cl)c([2H])c([2H])c2c1[2H]
IUPAC navn 2-chloro-1,3,4,5,6,7,8-heptadeuterionaphthalene
Molekylvægt (g/mol) 169.66
Formel vægt 169.0676